missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce Plant Total Protein Extraction Kit

The Thermo Scientific Pierce Plant Total Protein Extraction Kit effectively extracts protein from all kinds of dry and fresh plant tissue without liquid nitrogen or organic solvents.

Brand:  Thermo Scientific™ A44056

203.77 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 16337746

  • 287.00 € / Each
Estimated Shipment: 05-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Features of the Plant Total Protein Extraction Kit:
Efficient—obtain excellent protein yields in less than 10 minutes using a single buffer and a spin column
Versatile—native or denaturing lysis buffers for protein extraction from leaves, stems, and seeds
Compatible—extracts can be quantified using the BCA Protein Assay Kit or the Rapid Gold BCA Protein Assay Kit

The Pierce Plant Total Protein Extraction Kit is composed of optimized buffers and filter cartridges that allow for efficient and rapid protein extraction in less than 10 minutes. The kit is designed to rapidly extract denatured or native proteins from plant tissues (leaves, seeds, soft stem, roots, etc.). Simply grind the sample in the lysis buffer in the filter cartridge and centrifuge. The protein extracts can be used for applications such as SDS-PAGE, western blotting, immunoprecipitation, affinity purification, and activity assays.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Western Blot
Room Temperature
50 preps
  • Denaturing Lysis Buffer, 25 mL
  • Native Lysis Buffer, 25 mL
  • Protein Extraction Filter Cartridges, 50 Ea.
  • Collection Tubes, 50 Ea.
  • Plastic Rod
Pierce™
Plant Total Protein Extraction Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.