missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ EZ-Link™ Plus Activated Peroxidase
Shop All Thermo Scientific Products
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mg
5 x 1mg
missing translation for 'unitSize'
1mg
Pack of 5
Description
Thermo Scientific™ EZ-Link™ Plus Activated Peroxidase Kit contains horseradish peroxidase enzyme that has been aldehyde-activated with periodate to enable high-efficiency labeling and preparation of protein or antibody conjugates.
Other amine-reactive chemistries, like glutaraldehyde, have a tendency to polymerize and produce lower amounts of viable conjugate. Sugar residues present on HRP have been oxidized with periodate to produce aldehydes that react with primary amines. Plus Activated Peroxidase is mixed with the protein to be coupled at a pH compatible with your protein or antibody. After incubating the reaction for one to two hours, the conjugate linkage is reduced and the activated HRP quenched with ethanolamine. The conjugate is purified by desalting or by dialysis. If you are preparing an antibody-HRP conjugate, use the Thermo Scientific Pierce Conjugate Purification Kit (Part No. 44920) to separate unreacted enzyme from the conjugate.
Highlights:
- Reacts with readily accessible primary amines to form a covalent secondary amine bond
- Can be stored for at least 12 months at -20°C
- Consistent conjugation yields reliable conjugates
- Enzyme activity is 120-200 units/mg
Specifications
Specifications
| Chemical Reactivity | Amine |
| Quantity | 1 mg |
| Reactive Moiety | Aldehyde |
| Solubility | Water |
| Content And Storage | Note: EZ-Link Plus Activated Peroxidase contains 1-3 moles of aldehyde per mole of peroxidase. Storage: Upon receipt store product protected from moisture at -20°C. Product is shipped with an ice pack. |
| Form | Solid |
| Product Line | EZ-Link |
| Label Type | Enzyme Labeled |
| Label or Dye | HRP (Horseradish Peroxidase) |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction