missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Premium Grade Sulfo-NHS (N-hydroxysulfosuccinimide)

Optimize applications where product integrity and risk minimization are paramount with our highest quality formulation of this N-hydroxysuccinimide modification reagent.

Brand:  Thermo Scientific™ PG82071


 View more versions of this product

Product Code. 15215763

  • 264.00 € / 500mg
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Optimize applications where product integrity and risk minimization are paramount with the specially characterized Thermo Scientific™ Pierce™ Premium Grade Sulfo-NHS, our highest quality formulation of this N-hydroxysuccinimide chemical modification reagent.

Thermo Scientific Pierce Premium Grade Sulfo-NHS is our highest quality formulation of this N-hydroxysuccinimide chemical modification reagent, specially characterized for applications where product integrity and risk minimization are paramount.

Compared to the standard grade product, Premium Grade Sulfo-NHS provides more clearly defined quality and product support by including (a) increased analytical testing and product characterization, (b) greater batch-specific information and quality assurance review, (c) extensive lot sample retention and (d) change control notification. Sulfo-NHS (N-hydroxysulfosuccinimide) enables control and modification of carbodiimide crosslinking reactions involving activation of carboxylates (–COOH) for conjugation with primary amines (–NH2). Derivatives are easily synthesized by mixing the sulfo-NHS with a carboxyl-containing molecule and a dehydrating agent such as the carbodiimide EDC (EDAC). The method is the basis for generating many types of protein labeling reagents, including amine-reactive fluorescent dyes, biotin affinity tags and pegylation compounds.

Highlights:
  • Sulfo-NHS – use with EDC crosslinker to prepare amine-reactive NHS ester reagents
  • High quality – identity and purity confirmed by several tests, including quantitative NMR
  • Product integrity – enhanced level of testing and characterization compared to standard grade
  • Lot retention – ample supply of past lots retained to ensure future process testing
  • Change management – Change Control Notification (CCN) service
  • Consistent manufacture – batch-specific manufacturing documentation review
Applications:
  • Improve efficiency of EDC coupling reactions
  • Convert carboxyls to amine-reactive sulfo-NHS esters
  • Membrane-impermable for cell surface labeling
  • Crosslink proteins to carboxyl-coated beads or surfaces more efficiently
  • Activate nanoparticles with amine-reactive sulfo-NHS esters
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Amine-Carboxyl
217.13
Upon receipt store at 4°C.
Ambient
Chemical Labeling
Carbodiimide
Powder
Water
Yes
No
No
No
Pierce™
Heterobifunctional
Short (<10 Å)
500 mg
Standard, Single-use, Premium-grade
Crosslinker
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Pierce Premium Grade Sulfo-NHS (N-hydroxysulfosuccinimide)

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.