Learn More
Thermo Scientific™ Pierce Protease Inhibitor Tablets
Thermo Scientific™ Pierce Protease Inhibitor Tablets and Mini Tablets are broad-spectrum protease inhibitors that are highly effective at preventing proteolytic degradation for cell lysis and protein extraction experiments.
Brand: Thermo Scientific™ A32963
243.34 EUR valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
Thermo Scientific™ Pierce Protease Inhibitor Tablets and Mini Tablets are broad-spectrum protease inhibitors that are highly effective at preventing proteolytic degradation for cell lysis and protein extraction experiments.
Additional Information and Online Ordering
Pierce Protease Inhibitor Tablets protect proteins from proteolytic degradation through inhibition of the serine-proteases, cysteine-proteases, aspartic acid-proteases and aminopeptidases that are typically present in cellular lysate samples. The tablets contain AEBSF, aprotinin, bestatin, E-64, leupeptin and pepstatin A and are available in formulations with or without EDTA, a metalloproteinase inhibitor. The EDTA-free formulation has added compatibility with IMAC purification methods and 2D gel electrophoresis. Stable for storage at 4°C for up to 12 months, each large tablet is sufficient for 50mL of solution and each mini tablet is sufficient for 10mL of solution. Both sizes and formulations of tablet are suitable for reagent preparation prior to protein extraction from tissue and cultured cells.
Highlights:
- Economical −both large and mini tablets are most cost effective than other commercially available formulations for equivalent volumes
- Easy−to−use − the refrigerator−stable tablets are easily dissolved in buffers and lysis reagents
- Two formulation − available with or without EDTA
- Compatible − use with homebrew or commercially available cell lysis reagents and Thermo Scientific Protein Assays
Specifications
Pierce Protease Inhibitor Tablets | |
1 Vial, Store at 2°C to 8°C | |
Solid | |
50 mL Solution | |
20 Tablets |
Protease Inhibitor | |
Room Temperature | |
Pierce™ | |
Protease Inhibitor |
For Research Use Only. Not for use in diagnostic procedures.