Learn More
Thermo Scientific™ Pierce™ Protease Inhibitor Tablets, EDTA-free
Thermo Scientific™ Pierce Protease Inhibitor Tablets are broad-spectrum protease inhibitors that are highly effective at preventing proteolytic degradation for cell lysis and protein extraction experiments
20.73 € - 362.00 €
Alert:
For Research Use Only. Not for use in diagnostic procedures.
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16346532
|
Thermo Scientific™
A32966 |
2 Tablets |
25.34 €
Each |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||||
15677308
|
Thermo Scientific™
A32965 |
20 Tablets |
362.00 €
Pack of 20 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16336532
|
Thermo Scientific™
A32964 |
2 Tablets | N/A | ||||||
Description
Pierce Protease Inhibitor Tablets protect proteins from proteolytic degradation through inhibition of the serine-proteases, cysteine-proteases, aspartic acid-proteases and aminopeptidases that are typically present in cellular lysate samples. The tablets contain AEBSF, aprotinin, bestatin, E-64, leupeptin and pepstatin A. Stable for storage at 4°C for up to 12 months, each mini or large tablet tablet is sufficient for 10mL or 50mL of solution, respectively, and is suitable for reagent preparation prior to protein extraction from tissue and cultured cells.Highlights:
- Improved formulation—tablets dissolve quickly into a clear solution
- Economical – more cost-effective than other commercially available formulations for equivalent volumes
- Convenient—ready-to-use, fully disclosed, broad-spectrum formulations
- Compatible—use directly with Thermo Scientific™ Pierce™ BCA Assays