Learn More
Thermo Scientific™ Pepstatin A Protease Inhibitor
Customize your protease inhibitor cocktail formulation with these small packages of individual protease inhibitor peptides and compounds.
Brand:  Thermo Scientific™ 78436
Product Code. 10036263
  • 246.00 € / 25mg
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Thermo Scientific™ Protease Inhibitors are small packages of individual protease inhibitor peptides and compounds for customized formulation or modification of protease inhibitor cocktails.
The individual reagents include AEBSF, aprotinin, bestatin, E64, leupeptin, pepstatin A, and PMSF. Although Thermo Scientific Halt™ Protease Inhibitor Cocktails provide convenient and optimized broad-spectrum protease inhibition for routine cell lysis and assay needs, certain applications require a more customized approach. By offering the individual high-quality reagents used to make our cocktail products, we provide you with the tools needed to apply a protease inhibitor individually, supplement a cocktail, or combine several components to make customized protease inhibitor cocktails for specialized applications.
Highlights:
  • High-quality reagents in convenient and affordable package sizes
  • Purchase individually to prepare customized protease inhibitor cocktails
  • Supplement Halt Protease Inhibitor Single-Use Cocktails, whose formulations are fully disclosed, to achieve desired concentrations of specific component reagents
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pierce Pepstatin A Protease Inhibitor
Store between 2°C and 8°C
Pierce™
Protease Inhibitor
Protease Inhibitor
Lyophilized
Protease Inhibitor
25 mg