Learn More
Thermo Scientific™ Aprotinin Protease Inhibitor
Customize your protease inhibitor cocktail formulation with these small packages of individual protease inhibitor peptides and compounds.
Brand: Thermo Scientific™ 78432
191.90 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
Thermo Scientific™ Protease Inhibitors are small packages of individual protease inhibitor peptides and compounds for customized formulation or modification of protease inhibitor cocktails.
The individual reagents include AEBSF, aprotinin, bestatin, E64, leupeptin, pepstatin A, and PMSF. Although Thermo Scientific Halt™ Protease Inhibitor Cocktails provide convenient and optimized broad-spectrum protease inhibition for routine cell lysis and assay needs, certain applications require a more customized approach. By offering the individual high-quality reagents used to make our cocktail products, we provide you with the tools needed to apply a protease inhibitor individually, supplement a cocktail, or combine several components to make customized protease inhibitor cocktails for specialized applications.
Highlights:
- High-quality reagents in convenient and affordable package sizes
- Purchase individually to prepare customized protease inhibitor cocktails
- Supplement Halt Protease Inhibitor Single-Use Cocktails, whose formulations are fully disclosed, to achieve desired concentrations of specific component reagents

Specifications
Pierce Aprotinin Protease Inhibitor | |
Store between 2°C and 8°C | |
Pierce™ | |
Protease Inhibitor |
Protease Inhibitor | |
Lyophilized | |
Protease Inhibitor | |
25 mg |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.