missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Protein A UltraLink™ Resin

Extremely durable polyacrylamide support with immobilized Protein A for standard-capacity IgG purification.

Brand:  Thermo Scientific™ 53139

Product Code. 11879400

  • 626.00 € / 5mL
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Protein A UltraLink Resin is an extremely durable beaded polyacrylamide support to which IgG-binding Protein A has been immobilized for use in IgG purification methods.

The Protein A UltraLink Resin is used to bind and purify IgG in gravity-flow, spin-column or medium-pressure chromatography methods. Protein A is a popular antibody-binding protein that is especially useful for affinity purification of IgG from rabbit or human serum.

UltraLink Resin is an azlactone-activated polyacrylamide support that is hydrophilic, charge-free, high capacity, highly crosslinked, rigid and porous. This support is especially useful for medium pressure techniques when using large sample volumes requiring fast-flow techniques (FPLC) and large-scale applications. Unlike typical beaded agarose supports, UltraLink Support does not crush or collapse when subjected to high flow rates that result in pressures exceeding 25psi.

Highlights:

  • Immobilized Protein A – specifically binds IgG from several mammalian species, especially most subtypes of human and rabbit IgG

  • Standard capacity – loading of immobilized Protein A provides a binding capacity of approx. 16mg human IgG per milliliter of resin

  • UltraLink Resin – acrylamide-based support provides an alternative to crosslinked beaded agarose supports for medium pressure applications

Properties of Thermo Scientific UltraLink Resin:

  • Support pH Stability: 1 to 13
  • Average Particle Size: 50 to 80 microns
  • Exclusion Limit: greater than 2,000,000 daltons
  • Average Surface Area: greater than 250m2 per gram of beads
  • Average Pore Volume: greater than 1.2mL per gram of beads (> 60% of bead volume)
  • Pore Size: 500 to 1000 angstroms (50 to 100nm)
  • Maximum Linear Velocity: 3000cm per hour
  • Maximum Pressure: 100psi (6.9 bar), defined as the maximum pressure drop across a column that the resin can withstand (Note: The indicated gauge pressure of a liquid chromatography apparatus may be measuring the total system pressure rather than the pressure drop across the column.)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Resin
UltraLink Affinity Resin
Protein A
Upon receipt store products at 4°C. Products are shipped at ambient temperature.
Pierce™, UltraLink™
Bottle
IgG Antibodies
5 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.