Learn More
Thermo Scientific™ Pierce™ Protein Coated Plates
Shop All Thermo Scientific ProductsDescripción
These plates are uniformly and stably coated with one of four popular immunoglobilin-binding proteins (Protein A, Protein G, Protein A/G, or Protein L). They enable antibodies, or pre-established antibody-antigen complexes, to be captured to wells of the polystyrene plate for subsequent detection in plate readers. The strategy is useful for a variety of assay situations, but it is generally not suitable for sandwich-type ELISA.
Thermo Scientific Pierce Protein Coated Plates provide an alternative to direct, passive adsorption methods for immobilizing antibodies for ELISA and other plate-based assay techniques.
Highlights
• Retain antibody activity, which can be lost when antibodies are immobilized by passive adsorption
• Orient antibodies for maximum antigen-binding capacity
• Immobilize antibodies for plate assays without prior purification
• Ensure minimal variation (<5% well-to-well) from consistent coating
• Reduce nonspecific binding because plates are pre-blocked with SuperBlock Blocking Buffer
• Bind 2 to 5 pmol of antibody per well (based on tests with rabbit IgG)
Protein A/G Coated Plates
• Fusion protein containing four antibody-binding sites from Protein A and two from Protein G
• Binds to most IgG species because it combines the specificities of Protein A and Protein G
• Binds to Fc region of antibodies, ensuring optimal orientation
• Binds well using a wide range of pH
Protein A Coated Plates
• Binds strongly to IgG from human, rabbit, guinea pig, pig, dog and rhesus monkey
• Binds strongly to mouse IgG2a, IgG2b and IgG3
• Binds to Fc region of antibodies for optimal orientation
Protein G Coated Plates
• Binds strongly to IgG from many species including human, mouse, rabbit, sheep, and goat
• Binds only to IgG—no cross-reactivity with other antibody classes
• Binds to Fc region of antibodies for optimal orientation
Protein L Coated Plates
• Binds to all classes of immunoglobulins (IgG, IgM, IgA, IgE and IgD)
• Binds to the VL region of kappa light chains (human I, III, IV and Mouse I) without interfering with antigen-binding sites
• Binds to Fab fragments and ScFv
• Does not bind bovine, goat or sheep immunoglobulin
• Binds weakly to rabbit immunoglobulin
• Pre-blocked to reduce nonspecific binding
Especificaciones
Especificaciones
| Binding Property | Standard Capacity, Affinity |
| Format | Strip Plate |
| Content And Storage | Note: Plates are activated to 1 μL and supplied pre-blocked with Thermo Scientific SuperBlock Blocking Buffer. Use these plates for single-antibody assays only. In multiple antibody assays, such as sandwich ELISAs, the first antibody cannot block all binding sites and subsequent antibodies will bind to the plate, resulting in false positives. Upon receipt store plates at 4°C in unopened pouches. Place opened, unused plates in a resealable bag and store desiccated at 4°C. Plates are shipped at ambient temperature. |
| Detection Method | Colorimetric |
| Product Line | Pierce |
| Color | Clear |
| Product Type | Detection Plate, Immunoassay, ELISA |
| Material | Polystyrene |
| No. of Wells | 96 |
| Surface Treatment | Protein A |
| Mostrar más |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.