missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Protein-Free Blocking Buffer

Pierce Protein-Free Blocking Buffer is a non-protein compound that provides effective blocking for membrane-based and plate-based protein detection methods, resulting in extremely low background.

Brand:  Thermo Scientific™ 37572

178.92 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10229663

  • 252.00 € / 1 L

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Pierce Protein-Free Blocking Buffer is a non-protein compound that provides effective blocking for membrane-based and plate-based protein detection methods, resulting in extremely low background. It is supplied in a convenient, ready-to-use format.

Features of Protein-Free Blocking Buffer include:
Protein-free blocker—minimizes or eliminates cross-reactivity associated with protein-based blocking buffers
Applications—compatible with western blotting (membranes), ELISA (microplates), and arrays (coated glass slides)
Streptavidin-friendly—free of biotin; no interference with avidin-biotin detection systems
High-performance—optimized and validated in many protein methods to provide high signal-to-noise ratio (i.e., no quenching of specific binding and signal but eliminating nonspecific binding and background)

Traditional blocking buffers contain proteins that can cross-react in certain immunodetection systems, resulting in high background and reduced signal. Pierce Protein-Free Blocking Buffers are devoid of protein while remaining highly effective at blocking plates, membranes, and other surfaces for ELISA, western blotting, glass slide arrays, and other applications.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Protein-Free (PBS) Blocking Buffer, 1 L
1 L
ELISA, Western Blot, Arrays
1X
Non-protein compound
Blocking Buffer, protein-free
Liquid
Pierce™
PBS
Upon receipt store product at room temperature. After opening, store product at 4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers