missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PVDF Transfer Membranes, 0.45 μm, 1 Roll

Pierce PVDF Transfer Membranes are made of high-quality polyvinylidene difluoride and provide high binding capacity for proteins and nucleic acids for western, Southern, and northern blotting methods.

Brand:  Thermo Scientific™ 88518

453.15 EUR valid until 2025-12-31
Use promo code "24363" to get your promotional price.


Product Code. 10617354

  • 504.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Pierce PVDF Transfer Membranes are made of high-quality polyvinylidene difluoride and provide high binding capacity for proteins and nucleic acids for western, Southern, and northern blotting methods. The 0.45-μm pore size make these membranes well-suited for a wide range of western blotting conditions and ideal for transfer of proteins >10 kDa. For best results, preactivate the membrane with 100% alcohol.

Features include:
• Re-probe characteristics—high mechanical strength makes PVDF an excellent membrane for stripping and re-probing
• Durability—compatible with most organic solvents, acids, and mild bases
• Compatible with commonly used transfer conditions and detection methods such as staining, chemiluminescence, and radiolabeling
• High sensitivity—exceptional binding capacity makes PVDF the ultimate choice for low abundance proteins

TRUSTED_SUSTAINABILITY
Specifications

Specifications

3.75 m
Pierce PVDF Transfer Membrane, 0.45 μm, 3.75 m x 26.5 cm
1 Roll
PVDF
Store membranes flat at ambient temperature and away from chemical vapors. Some solvent vapors may partially dissolve the membranes, which will disrupt the pore structure.
3.75 m x 26.5 cm
26.5 cm

294 μg/cm2
BSA: 215 μg/cm2
Insulin: 160 μg/cm2

Roll
0.45 μm
Room Temperature
≥100 Mini-gel Blots (When cut to 8.8 x 10 cm); ≥80 Midi-gel Blots (When cut to 8.5 x 13.25 cm)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZL403309 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number88518,88518_LITL18 PROMO
Lot NumberZK397607 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number88518,88518_LITL18 PROMO
Lot NumberZI397606 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number88518,88518_LITL18 PROMO
Lot NumberZG396343A Certificate TypeCertificate of Analysis Date07/08/2024 Catalog Number88518,88518_LITL18 PROMO
Lot NumberZG396343 Certificate TypeCertificate of Analysis Date07/08/2024 Catalog Number88518,88518_LITL18 PROMO
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ PVDF Transfer Membranes, 0.45 μm, 1 Roll > 3.75 m x 26.5 cm, Roll, 0.45 μm, 1 Roll

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.