missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ Pyrogen-Free Glass Test Tubes
The endotoxin-free glass test tubes contain less than 0.005 EU/mL endotoxin. Recommended for use as reaction tubes in Gel Clot assays.
Brand: Thermo Scientific™ A43889
25.42 EUR valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
These endotoxin-free glass test tubes contain less than 0.005 EU/mL endotoxin. They are recommended for use as reaction tubes in endotoxin detection assays.Specifications
Endotoxin level <0.005 EU/mL | |
Gel Clot and Chromogenic Endotoxin Assay | |
Test Tube |
Glass | |
30 tubes |
For Research Use Only. Not for use in diagnostic procedures.