missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Quantitative Peroxide Assay Kit (Lipid)

Detect and measure hydrogen peroxide levels (H2O2) in biological samples using an iron (Fe) and xylenol orange (XO) reagent for microplates or tubes.

Brand:  Thermo Scientific™ 23285

Product Code. 10361494

  • 239.00 € / 1 set
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Detect and measure hydrogen peroxide levels in biological samples using an iron and xylenol orange reagent with Thermo Scientific™ Pierce Quantitative Peroxide Assay Kits.

The Thermo Scientific Pierce Quantitative Peroxide Assay Kit (lipid-compatible formulation) detects and measures hydrogen peroxide levels in biological samples using an iron and xylenol orange (XO) reagent.

This lipid-compatible peroxide assay kit detects peroxides based on oxidation of ferrous to ferric ion in the presence of xylenol orange. The kit's formulation may be used with lipid-containing samples without first extracting the lipids; however, because this kit does not contain sorbitol, the assay is less sensitive. Most proteins do not interfere with this assay, although some metal chelators may require use of a blank (i.e., control) to mitigate these effects.

Features of the Quantitative Peroxide Assay Kit:
Peroxides—detects and measures hydrogen peroxide (H2O2) levels in biological and other liquids samples
Convenient—no proprietary reagents involved but the ready-to-use kit removes the variability, difficulty and added expense of formulating the reagents yourself

Applications:
• General hydrogen peroxide (H2O2) measurement in protein samples
• Quantitation of peroxides in detergents
• Assessment of the effects of peroxides on cellular activity
• Determination of protein glycation; Measurement of peroxide accumulation in lipids

In this assay, hydroperoxides convert the Fe2+ to Fe3+ at acidic pH. With this lipid-compatible formulation, the peroxide converts the Fe2+ to Fe3+ directly. In a sulfuric acid solution, the Fe3+ complexes with the xylenol orange (XO) dye to yield a purple product having an absorbance maximum at 560 nm. Peroxide levels in test samples can be determined by calculation from the known extinction coefficient of the XO-Fe complex or by reference to a standard curve prepared with hydrogen peroxide solution.

Related Products
Pierce™ Quantitative Peroxide Assay Kit, Aqueous (Cat. No. 23280)
Pierce™ Peroxide Assay Reagent A (Cat. No. 23281)
Pierce™ Peroxide Assay Reagent B (Cat. No. 23282)
Pierce™ Peroxide Assay Reagent C (Cat. No. 23283)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sufficient For: 100 tube assays or 500 microplate assays with lipid-containing samples
• Reagent A, 1 mL
• Reagent C, 4 x 25 mL

Store at 4°C.
Quantitative Peroxide Assay Kit
Pierce™
Not Target-Specific
Solution-based Detection, Absorbance
100 mL
Pierce Quantitative Peroxide Assay Kit (Lipid)
Colorimetric
Quantitative Peroxide Assay
100 Tube Assays or 500 Microplate Assays
Spectrophotometer, Microplate Reader
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.