missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ Recombinant Protein L, Biotinylated
Shop All Thermo Scientific Products
Click to view available options
Quantity:
0.5 mg
Unit Size:
500µg
Description
Purified and conjugated forms of Thermo Scientific™ Pierce Recombinant Protein L enable probing and detection of mouse and human antibodies containing kappa light chains in Western blotting, ELISA, IHC and other immunoassay protocols.
Protein L is an immunoglobulin-binding protein that was originally derived from the bacteria Peptostreptococcus magnus, but is now produced recombinantly. Protein L has the unique ability to bind through kappa light chain interactions without interfering with an antibody's antigen-binding site. This gives Protein L the ability to bind a wider range of Ig classes and subclasses than other antibody-binding proteins. Protein L will bind to all classes of Ig (IgG, IgM, IgA, IgE and IgD). Protein L will also bind single chain variable fragments (scFv) and Fab fragments. Protein L binds kappa I, III, and IV in human and kappa I on mouse.
Highlights:
- Contains four immunoglobulin-binding domains per protein
- Binds members of all classes of Ig (IgG, IgM, IgA, IgE and IgD) that contain kappa light chains
- Binds single chain variable fragments (scFv) and Fab fragments that contain kappa light chains
Specifications
Specifications
| Product Type | Protein L, Biotinylated |
| Conjugate | Biotin |
| Quantity | 0.5 mg |
| Product Line | Pierce |
| Form | Powder |
| Protein Subtype | Protein L |
| Target | Antibodies |
| Content And Storage | Upon receipt store at -20°C. Product is shipped at ambient temperature. |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction