Learn More
Thermo Scientific™ Pierce™ SATP (N-succinimidyl-S-acetylthiopropionate)
Short (4.1A), amine-reactive (NHS ester) sulfhydryl-addition reagent; creates stable (protected yet exposable) -SH group for protein crosslinking strategies.
Brand:  Thermo Scientific™ 26100
Product Code. 10320974
  • 122.00 € / 50mg
Estimated Shipment: 02-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Thermo Scientific™ Pierce SATP is a variant (4.1 Å spacer arm) of SATA, a sulfhydryl-addition reagent for covalent modification of primary amines to facilitate controlled, heterobifunctional crosslinking.
Highlights:
  • Reacts with primary amines to add protected sulfhydryls
  • An easy deprotection step generates a free sulfhydryl group
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Amine
Sulfhydryl Group
Pierce™
Active Ester, Succinimidyl Ester, NHS Ester
Powder
50 mg
Crosslinker
Upon receipt store desiccated at -20°C. Product is shipped at ambient temperature.
Conjugating Groups
Heterobifunctional
Medium (10 to 30 Å)
SATP
DMF, DMSO