Learn More
Thermo Scientific™ Sodium Deoxycholate Detergent
Water-soluble, ionic detergent used in cell lysis buffers (e.g., RIPA buffer) to disrupt protein interactions for liposome and membrane protein isolation.
155.00 € - 564.00 €
Specifications
Product Type | Detergent |
---|---|
Content And Storage | Store at -20°C |
Form | Powder |
Reagent Type | Detergent (Pure) |
Description
Thermo Scientific™ Sodium Deoxycholate is an ionic detergent that is especially useful for disrupting and dissociating protein interactions.
Sodium deoxycholate (deoxycholic acid) is a water-soluble, bile-acid, ionic detergent commonly used in protein methods. It is most frequently used as a component of cell lysis buffers (e.g., RIPA buffer) but also has been used for liposome preparation, isolation of membrane proteins and lipids, preventing nonspecific binding in affinity chromatography and a cell culture media supplement.Highlights:
- Popular anionic, bile-acid detergent for many laboratory uses
- Effective in disrupting and dissociating many types of protein interaction
- Can be removed from solution by dialysis
- Useful for elution or regeneration of certain kinds of affinity columns
- High-purity compound with low UV absorptivity
Detergent Properties:
- Alternative Names: Sodium deoxycholic acid; deoxycholate, sodium salt
- Chemical Name: 3, 12-α-Dihydroxy-5β-cholan-24-oic acid, monosodium salt
- Molecular Weight: 414.6
- Detergent Class: Ionic (anionic)
- Aggregation Number: 5 (average)
- Micelle Molecular Weight: 2000 (average)
- Critical Micelle Concentration (CMC): 2 to 6mM (0.083 to 0.249%, w/v)
- Cloud Point: Unknown
- Dialyzable: Yes
Specifications
Detergent | |
Store at -20°C | |
Powder | |
Detergent (Pure) |
Safety and Handling
- Sodium Deoxycholate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash hands thoroughly after handling.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium 3-droxy-5-oate
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.