missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ High Sensitivity Streptavidin-HRP

Amplify signal in Western blot, ELISA or other biotin-based detection methods with this exclusive, high signal-to-noise, peroxidase-conjugated, ready-to-use streptavidin.

280.00 € - 1154.00 €

Specifications

Concentration 1.0–1.1 mg/mL
Conjugate HRP
Content And Storage Storage at 4°C for at least one year.
Product Line Pierce™
Target Molecule Biotin
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10598893
View Documents
Thermo Scientific™
21130
0.5 mL
280.00 €
0.50mL
In Stock
to see stock.
Add to basket
 
11896814
View Documents
Thermo Scientific™
21132
5 mL
1154.00 €
5mL
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
 
Description

Description

Thermo Scientific™ Pierce High Sensitivity Streptavidin HRP Conjugate is an exclusive peroxidase-conjugated streptavidin biotin-binding protein that provides signal amplification like poly-HRP and exceptional storage stability.

  • Thermo Scientific Pierce High Sensitivity Streptavidin-HRP is an exclusive peroxidase-conjugated streptavidin biotin-binding protein
  • It provides signal amplification like poly-HRP and exceptional storage stability
  • This specially-manufactured variety of HRP-conjugated streptavidin protein meets the demands of today's scientists for more sensitive detection in ELISA and Western blotting applications
  • Conjugate is suitable for use with chemiluminescent, chemifluorescent, or colorimetric substrates
  • Each High Sensitivity HRP conjugate is packaged in an easy-to-use stabilized solution that enables convenient storage at 4°C for at least one year
  • Experiments indicate that the High Sensitivity Streptavidin-HRP can be diluted 8-to 10-fold more than traditional HRP conjugates and still produce similar results
Specifications

Specifications

1.0–1.1 mg/mL
Storage at 4°C for at least one year.
Biotin
HRP
Pierce™
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.