missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Sulfo-SMCC (sulfosuccinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate), No-Weigh™ Format
Shop All Thermo Scientific Products
Click to view available options
Quantity:
50 mg
10 x 2 mg
1 g
Unit Size:
1g
50mg
Pack of 10
Descrizione
Thermo Scientific™ Pierce Sulfo-SMCC is a very popular, water-soluble, amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a medium-length cyclohexane spacer arm (8.3 Ås).
- Reactive groups: sulfo-NHS ester and maleimide
- Reactive towards: amino and sulfhydryl groups
- Water-soluble (compare to SMCC), so crosslinking can be done in physiologic solutions
- High purity, crystalline reagent can be used to create high-purity maleimide-activated derivatives
- Cyclohexane bridge confers added stability to the maleimide group making Sulfo-SMCC the ideal crosslinking agent for maleimide activation of proteins. Maleimide groups are stable for 64 hours in 0.1 M sodium phosphate buffer, pH 7 at 4°C.
Specifica
Specifica
| Chemical Reactivity | Amine-Sulfhydryl |
| Cleavable | No |
| Description | Sulfo-SMCC |
| Molecular Weight (g/mol) | 436.37 |
| PEGylated | No |
| Spacer Arm Length | 8.3 Å |
| Content And Storage | Upon receipt store desiccated at -20°C. |
| Cell Permeability | No |
| Shipping Condition | Ambient |
| Product Line | No-Weigh |
| Vedi altri risultati |
For Research Use Only. Not for use in diagnostic procedures.
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto