missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ TCEP-HCl, No-Weigh™ Format

Reduce protein and peptide disulfide bonds using this thiol-free, pure, crystalline tris (2-carboxyethyl) phosphine hydrochloride, TCEP (CAS 5961-85-3).

115.00 € - 638.00 €

Specifications

Form Solid
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10530434
View Documents
Thermo Scientific™
20490
1 g
115.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
11856784
View Documents
Thermo Scientific™
20491
10 g
638.00 €
10g
Please to purchase this item. Need a web account? Register with us today!
15780329
View Documents
Thermo Scientific™
A35349
10 x 1 mg
130.00 €
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Thermo Scientific™ Pierce TCEP-HCl is pure, crystalline Tris(2-carboxyethyl)phosphine hydrochloride, a thiol-free compound that is highly effective at reducing protein and peptide disulfide bonds, as well as being odorless and stable.

Highlights:

  • Reduces even the most stable water-soluble alkyl disulfides
  • Effective at room temperature and pH 5 in less than five minutes
  • Does not require removal before reaction with maleimide or haloacetyl compounds
  • Water solubility of 310g/L
  • Resistant to air oxidation; nonvolatile and nonreactive toward other functional groups in proteins
Specifications

Specifications

Solid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • TCEP HCl, Tris(2-Carboxyethyl) Phosphine Hydrochloride

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

Supplemental information
  • MIXTURE LIST-Contains : Tris(2-carboxyethyl)phosphonium chloride

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.