missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Trifluoroacetic Acid (TFA), Sequencing grade

Bottles of 99.5%-pure TFA (MW 114.02, CAS 76-05-01) liquid (sold by weight); for HPLC use as an ion-pairing agent in reverse-phase peptide separations.

153.00 € - 756.00 €

Specifications

Name Note Trifluoroacetic Acid (TFA)
Percent Purity 99.5% (TFA)
Physical Form Liquid
View More Specs

Products 8
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11808223
View Documents
Thermo Scientific™
TS-28904
10 x 1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10270594
View Documents
Thermo Scientific™
28904
10 x 1 mL
189.00 €
10mL
Please to purchase this item. Need a web account? Register with us today!
10762735
View Documents
Thermo Scientific™
28902
10 x 1 g
156.00 €
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
11501084
View Documents
Thermo Scientific™
TS-28902
10 x 1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10628494
View Documents
Thermo Scientific™
28903
100 g
239.00 €
100g
Please to purchase this item. Need a web account? Register with us today!
11511084
View Documents
Thermo Scientific™
TS-28903
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
13416806
View Documents
Thermo Scientific™
TS-28901
500 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11875825
View Documents
Thermo Scientific™
28901
500 mL
756.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Thermo Scientific™ Pierce Trifluoroacetic Acid (TFA) is manufactured and tested to meet strict specifications that ensure superior performance for use as an ion-pairing agent in reverse-phase peptide separations.

For complex peptide separations, the key to success can be to vary selectivity. Varying mobile phase composition on the same column can change selectivity enough to resolve peptide that would otherwise overlap. The TFA concentration is usually specified as 0.1% for reverse-phase HPLC of peptides. For reproducible separations from run-to-run or from lab-to-lab, it is essential to make concentrations the same.

Highlights:

  • Purity: >99.5% TFA and exceptional clarity allows sensitive, nondestructive peptide detection at low UV wavelengths
  • High-performance packaging: Packaged under nitrogen in amber glass with protective TFE-lined fluorocarbon caps to ensure TFA integrity
  • Choice of formats for convenience: 1 mL ampules can prepare 1 L of 0.1% v/v TFA solution for the mobile phase in reverse-phase chromatography in moments
  • CAS: 76-05-1

Storage Information

  • Store in original container protected from direct sunlight in a dry, cool and well-ventilated area
  • Should be stored from 20° to 25°C

Order Info

X in the ordering table indicates that hazardous shipping charges apply.

Specifications

Specifications

Trifluoroacetic Acid (TFA)
99.5% (TFA)
Liquid
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C.

Product Identifier
  • Trifluoroacetic Acid

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P315-Get immediate medical advice/attention.
  • P305-IF IN EYES:.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.