Learn More
Thermo Scientific™ Vacuum Hydrolysis Tube, 18 mL
High-quality valved glass tubes designed for heated, vacuum, acid-hydrolysis of proteins and peptides for amino acid analysis by HPLC chromatography.
Brand: Thermo Scientific™ 29572
Description
Thermo Scientific™ Pierce Vacuum Hydrolysis Tubes are high-quality glass tubes designed for heated, acid-based, vacuum hydrolysis of proteins and peptides for amino acid analysis.
Highlights:
- Upper temperature limit of the Vacuum Hydrolysis Tubes is 260°C; however, do not heat the tubes greater than 110°C in an oven Vacuum hydrolysis tubes fit conveniently into Reacti-Block™ Aluminum Heating Blocks

Specifications
Vacuum Hydrolysis Tube, 18mL | |
0.0 | |
1 Tube |
Vacuum Hydrolysis Tube | |
Tube |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.