Learn More
Thermo Scientific™ Piko PCR Plate, 24-well, clear
Thermo Scientific™ Piko PCR Plates are modular, ultra-thin wall PCR plates for Piko* Thermal Cyclers.
Brand: Thermo Scientific™ SPL024050
This item is not returnable.
View return policy
Description
Thermo Scientific™ Piko™ PCR Plates are modular, ultra-thin wall PCR plates for Piko Thermal Cyclers.
>Highlights• Ultra Thin Wall (UTW) for fast PCR/qPCR applications
• Low profile
• Designed for use with the Piko and PikoReal 24-well thermal cyclers
• Plates can be snapped into the plate frame to create a standard 96-well plate
• Compatible with standard multi-channel pipettes and liquid-handling platforms
• Well spacing and footprint conform to industry (ANSI) dimensions
Applications
• PCR
Related Products
Piko PCR Plate, 24-well, white
Piko PCR Plate, 24-well, clear
Plate Frame for 24-well Piko PCR Plates, white
Specifications
No Barcode | |
50 Plate | |
Clear | |
Piko Thermal Cycler, PikoReal Real-Time PCR System | |
24 | |
Low profile |
Certified RNase/DNase-free | |
Piko | |
PCR Microplate | |
Polypropylene | |
50 plates |