Learn More
Thermo Scientific™ Piko PCR Plate, 24-well, clear
Thermo Scientific™ Piko PCR Plates are modular, ultra-thin wall PCR plates for Piko* Thermal Cyclers.
Brand:  Thermo Scientific™ SPL0240
 View more versions of this product
Product Code. 10036339
  • 458.00 € / Pack of 200
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Thermo Scientific™ Piko™ PCR Plates are modular, ultra-thin wall PCR plates for Piko Thermal Cyclers.
Piko PCR Plates are also flexible alternatives for conventional microplates. Four plates, each the size of a microscope slide, can be reversibly snapped into a rigid frame, producing the equivalent of a standard 96- or 384-well microplate. This format allows multiple sample sources to be combined into a single PCR run, or subjected to different post-PCR processing.
  • Wall thickness half that of conventional thin-walled tubes
  • Flexible format saves cost and reduces waste
  • Compatible with standard multi-channel pipettes and liquid handlers
  • Well spacing and footprint correspond to industry (ANSI) dimensions
  • Available in clear and white
Recommended for:
PCR, Fast PCR, High Performance PCR
TRUSTED_SUSTAINABILITY
Specifications
Specifications
No Barcode
200 Plate
Clear
Piko Thermal Cycler, PikoReal Real-Time PCR System
24
Low profile
Certified RNase/DNase-free
Piko
PCR Microplate
Polypropylene
200 plates