Learn More
OHAUS™ Pioneer™ Precision Balance
Affordable balance to achieve reliable results
Brand:  OHAUS™ 30619384
1219.06 EUR valid until 2025-12-31
Use promo code "24377" to get your promotional price.
 View more versions of this product
Product Code. 17268770
  • 1295.00 € / 1 unit
Explore more special offers
Description
Description
  • Offers high accuracy and repeatability for essential weighing applications in laboratory, industrial and education settings at an economical price point
  • Featuring cast metal lower housing, sub-pan and stainless steel weighing pan
    • Durably constructed for versatile, long-term use
  • Second line display for additional information/guidance and USB/RS232 connectivity for easy communication
    • draft shield models features static removal (ESR) bar for convenient grounding
Key Features:
  • User-selectable environmental filters and brightness settings
  • Auto-tare
  • Auto-dim
  • User-selectable span calibration points
  • Software lockout and reset menu
  • User selectable communication settings and data print options
  • User-definable project and user IDs
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RS232 and USB device (included), GLP/GMP data output with real-time clock
Backlit LCD
Basic weighing, Parts counting, Percent weighing, Animal weighing, Density determination
Metal base, Plastic top housing, Removable stainless steel pan, Glass draft shield
0.01 g
50 to 86 °F, Operability guaranteed between 41 to 104 °F
Analytical and precision balances
321 x 209 x 98 mm
209 mm
180 mm
3.5 kg
6200 g
0.01 g
0.02 g
PX6202/E
1 sec.
10 to 30 °C, Operability guaranteed between 5 to 40 °C
Gram, Kilogram, Ounce, Pound, Carat, Pennyweight, Ounce Troy, Grain, Newton, Hong Kong Tael, Singapore Tael, Taiwan Tael, Momme, Tical, Mesghal, Tola, Baht, Custom
321 mm
98 mm
7.7 lb.