Learn More
Ohaus™ Pioneer™ PX 124M Analytical Balance
The Pioneer PX combines essential weighing functionality with competitive performance, offering high accuracy and repeatability for applications in laboratory, industrial and education settings.
Brand:  Ohaus™ 30430010
2045.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15805347
  • 2100.00 € / Each
Explore more special offers
Description
Description
  • Competitive Performance at an Economical Price - The PX offers high accuracy and repeatability for essential weighing applications in laboratory, industrial and educational settings at an economical pricepoint.
  • Durably Constructed for Long-Term Use - Featuring a cast metal lower housing, sub-pan and stainless steel weighing pan, the PX is durably constructed for versatile, long-term use.
  • Designed for Simple, Intelligent Operation - The PX features a second line display for additional information or guidance, a static removal bar for convenient grounding, and USB and RS232 connectivity for easy communication.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
InCal™ - Semi Automatic
RS232 & USB
120 g
0.000100 g
0.000200 g
PX124M Approved
3.000 sec.
321 x 209 x 309 mm
321.00 mm
309.000 mm
90 mm
7 kg
CE
Electronic Balance, PX124M
Backlit LCD
English (US), French, German, Italian, Spanish (MX)
ABS Plastic, Cast Metal, Stainless Steel, Glass
0.000100 g
Basic Weighing, Parts Counting, Percent Weighing, Animal Weighing, Density Determination
321.00 x 209.00 x 309.000 mm
209.00 mm
321.00 mm
4.5 kg
100/240 V ∼200 mA 50/60 Hz 12/18 VA