Learn More
Brady™ Pipe Banding Tape
Solid color pipe banding tape provides 360 degree visibility to improve safety and operational efficiency.
Brand:  Brady™ 275200
 View more versions of this product
Product Code. 12759884
  • 62.15 € / 1 roll
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Solid color pipe banding tape provides 360 degree visibility to improve safety and operational efficiency.
Specifications
Specifications
ROHS Environmental Compliance, EU Directive 2002/95/EC, CSA Accepted, UL Recognized,
Pipe Banding Tape
Yellow
Tape
Thermal Transfer
Pipe and valve marking
1 Ea.
-80°C to +110°C
0.071 mm
108.27 ft.
0.98 in.
PIPEBANDINGTAPEYELLOW25MMX33M
Permanent Acrylic, Permanent
Satin
Polyester
Pipe Marker
1 Roll
-112°F to +230°F
0.0028 in.
33 m x 25 mm
33 m
25 mm