missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PiPer™ Pyrophosphate or Phosphate Assay Kit

PiPer Pyrophosphate Assay Kit provides reagents for fluorogenic or colorimetric detection as low as 0.4 mμM (fluorescence) or 2 μM (absorbance) inorganic pyrophosphate (PPi) while Phosphate Assay kits can detect down to 0.2 μM Pi in purified enzyme systems. Each kit has enough for 1000 labelings.

754.00 € - 768.00 €

Specifications

Color Red
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Colorimetric, Fluorescence
For Use With (Equipment) Spectrophotometer, Microplate Reader
Dye Type Amplex™ Red
View More Specs

Products 2
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
10504053
View Documents
Invitrogen™
P22061
Phosphate Assay
768.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
10378442
View Documents
Invitrogen™
P22062
Pyrophosphate Assay
754.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PiPer Pyrophosphate and Phosphate Assay Kit provides an ultrasensitive assay to detect the fluorescent product resorufin. These kits can be used to detect inorganic pyrophosphate (PPi) or phosphate (Pi) in a variety of samples or can monitor the kinetics of their release by a variety of enzymes, including DNA/RNA polymerases, adenylate cyclase, S-acetyl coenzyme A synthetase, ATPases, GTPases, 5′-nucleotidase, protein phosphatases, acid/alkaline phosphatases and phosphorylase kinase. As resorufin has a strong absorption, this assay can be performed either fluorometrically or colorimetrically.

Key Features:

  • High Sensitivity: Can detect low levels of inorganic phosphate
  • Number of labelings per kit: ~1,000 of 100 μL per assay volume
  • Detection method: Colorimetric, Fluorescence
  • Compatibility: Can be used with both microplate readers and spectrophotometers
  • Excitation/emission maxima: 563/587 nm
  • Versatility: Suitable for a wide range of applications including enzyme activity assays
  • Quantitative Results: Provides accurate and reproducible results.

The PiPer Pyrophosphate Assay Kit provides a sensitive fluorogenic or colorimetric method for detecting as little as 0.4 μM (fluorescence) or 2 μM (absorbance) inorganic pyrophosphate in a 100 μL assay volume. The PiPer Phosphate Assay Kit detects as little as 0.2 μM Pi (fluorescence) in purified enzyme systems with a 100 μL assay volume.

Use the PiPer Pyrophosphate and Phosphate Assay Kit for applications such as: Anion Detection, Biochemical Toxicology Assays, Cell Analysis, Cell Metabolism, Cell Viability, Cellular Toxicology Assays, Drug Discovery and Development, Enzyme and Protein Activity Assays, Ionic Homeostasis and Signaling, Phosphatase Activity, Phosphatase Activity Assays, Phosphatase Biology, Target and Lead Identification and Validation.

The absorption and fluorescence of resorufin are pH dependent. There is a pKa at ~6.0 where the absorption maximum shifts to ~480 nm and the fluorescence quantum yield is markedly lower. Given Amplex™ Red reagent is unstable at high pH (>8.5) and is sensitive to light, we recommend this reaction be performed using the provided reaction buffer (at optimal pH ~7.5) for best results and reagents kept protected from light. All kits should follow user manual guidelines.

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Red
Colorimetric, Fluorescence
Amplex™ Red
PiPer™
Room Temperature
Store in freezer -5°C to -30°C and protect from light.
Spectrophotometer, Microplate Reader
96-well plate
1000 Assays
563/587 nm
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.