Learn More
Piperacetazine, MedChemExpress
MedChemExpress Piperacetazine is an antipsychotic prodrug, used forschizophrenia.
Brand:  Medchem Express HY-B1152
Encompass_Preferred
 View more versions of this product
Product Code. 16461098
  • 97.20 € / 5mg
Explore more special offers
Description
Description
Piperacetazine is an antipsychotic prodrug, used forschizophrenia.
Specifications
Specifications
Piperacetazine
Yellow
C24H30N2O2S
Ethanol : 50 mg/mL (121.78 mM; Need ultrasonic) ∣DMSO : ≥ 6 mg/mL (14.61 mM)
410.57
99.03%
Solid
3819-00-9
Research
5 mg
CC(C(C=C1N2CCCN3CCC(CCO)CC3)=CC=C1SC4=C2C=CC=C4)=O
410.57
Research
Neuroscience-Neuromodulation