missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piperazine hydrate, ≥97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1000 g
5000 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 42°C to 46°C |
Color | White |
pH | 11 to 12 |
Boiling Point | 125°C to 130°C |
Flash Point | >100°C (212°F) |
Odor | Weak, characteristic |
Quantity | 250 g |
UN Number | UN2579 |
Formula Weight | 86.14 (Anhydrous) |
Percent Purity | ≥97% |
Show More |
Safety and Handling
Product Identifier
- Piperazine hydrate
- Danger
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Piperazine
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'