missing translation for 'onlineSavingsMsg'
Learn More

Piperidine, 99%, extra pure

Product Code. 10159831 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
1 L
10 L
Unit Size:
1 L
10 L
100mL
500mL
This item is not returnable. View return policy

Product Code. 10159831

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-89-4 | C5H11N | 85.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-89-4
Molecular Formula C5H11N
Molecular Weight (g/mol) 85.15
InChI Key NQRYJNQNLNOLGT-UHFFFAOYSA-N
IUPAC Name piperidine
SMILES C1CCNCC1

Specifications

Melting Point -11.0°C
Density 0.8620 g/mL
Boiling Point 106.0°C
Flash Point 16°C
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4515 to 1.4535
Quantity 100 mL
Beilstein 20, 6
Fieser 01,886; 02,332; 04,393; 06,472; 07,293
Merck Index 15, 7580
Specific Gravity 0.862
Solubility Information Solubility aq. soln.: miscible. Other solubilities: soluble in alcohol and benzene, soluble in chloroform, acetone and ethanol
Viscosity 1.46 mPa.s (20°C)
Formula Weight 85.15
Grade Extra Pure
Chemical Name or Material Piperidine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Piperidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.