missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piperonyl butoxide, tech. 90%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
100 mL
Description
Piperonyl butoxide is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Piperonyl butoxide is used as a pharmaceutical intermediate.
Solubility
Soluble in benzene, ethanol and methanol. Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Piperonyl butoxide is used as a pharmaceutical intermediate.
Solubility
Soluble in benzene, ethanol and methanol. Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Specifications
Density | 1.059 |
Boiling Point | 155°C (0.3 mmHg) |
Flash Point | 171°C (340°F) |
Refractive Index | 1.498 |
Quantity | 100 mL |
UN Number | UN3082 |
Beilstein | 288063 |
Solubility Information | Soluble in benzene,ethanol and methanol. Not miscible or difficult to mix with water. |
IUPAC Name | 5-[2-(2-butoxyethoxy)ethoxymethyl]-6-propyl-1,3-benzodioxole |
Formula Weight | 338.44 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS09_Aquatic_Toxicity.png)
Product Identifier
- Piperonyl butoxide
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Piperonyl butoxide
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction