missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals PIPES, 98.5+%, for biochemistry
38.09 € - 1372.83 €
Chemical Identifiers
CAS | 5625-37-6 |
---|---|
Molecular Formula | C8H18N2O6S2 |
Molecular Weight (g/mol) | 302.36 |
InChI Key | IHPYMWDTONKSCO-UHFFFAOYSA-N |
Synonym | 1, 4-Piperazinebis(ethanesulfonic acid) |
PubChem CID | 79723 |
ChEBI | CHEBI:44933 |
IUPAC Name | 2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid |
SMILES | C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10711261
|
Thermo Scientific Chemicals
172610250 |
25 g | Plastic bottle |
38.09 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10062200
|
Thermo Scientific Chemicals
172611000 |
100 g | Plastic bottle |
102.51 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10731414
|
Thermo Scientific Chemicals
172615000 |
500 g | Plastic bottle |
352.76 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11956521
|
Thermo Scientific Chemicals
172610025 |
2.5 kg | Plastic bottle |
1372.83 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
5625-37-6 | |
302.36 | |
1, 4-Piperazinebis(ethanesulfonic acid) | |
CHEBI:44933 | |
C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O |
C8H18N2O6S2 | |
IHPYMWDTONKSCO-UHFFFAOYSA-N | |
79723 | |
2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid |
Specifications
(0.1M in 0.1M NaOH) (1 cm cell), (0.1M in 0.1M NaOH) (1 cm cell) at 0nm, 0.04 max. at 280nm, 0.05 max. at 260nm | |
5625-37-6 | |
Micro-crystalline Powder | |
Authentic | |
C8H18N2O6S2 | |
IHPYMWDTONKSCO-UHFFFAOYSA-N | |
2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid | |
79723 | |
302.35 | |
Biochemical |
(0.1M in 0.1M NaOH) (1cm cell) | |
White | |
98.5+% | |
0.5% max. | |
1, 4-Piperazinebis(ethanesulfonic acid) | |
C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O | |
302.36 | |
CHEBI:44933 | |
≥98.50% | |
PIPES |
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title