missing translation for 'onlineSavingsMsg'
Learn More

PIPES, disodium salt, 99%, Thermo Scientific Chemicals

Product Code. 11986521
Click to view available options
Quantity:
25 g
100 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76836-02-7
Molecular Formula C8H16N2Na2O6S2
Molecular Weight (g/mol) 346.324
InChI Key GMHSTJRPSVFLMT-UHFFFAOYSA-L
Synonym 1, 4-Piperazinediethanesulfonic acid disodium salt
PubChem CID 173553
ChEBI CHEBI:63055
IUPAC Name disodium;2-[4-(2-sulfonatoethyl)piperazin-1-yl]ethanesulfonate
SMILES C1CN(CCN1CCS(=O)(=O)[O-])CCS(=O)(=O)[O-].[Na+].[Na+]

Specifications

Absorbance 0.050 max. (0.1M in water) at 260nm, 0.050 max. (0.1M in water) at 280nm, 0.050 max. (260nm, 0.1M in water)
Color White
pH 9.2 to 10.0 max. (25°C, 1% min. aq. soln.)
Physical Form Crystalline Powder or Powder
Quantity 100 g
Assay Percent Range 99%
Infrared Spectrum Authentic
Packaging Glass bottle
Formula Weight 346.33
Percent Purity 99%
Chemical Name or Material PIPES, disodium salt
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'