Learn More
Sartorius Pipet Charging Stand and Carousel
Saves bench space. Sartorius Pipet Charging Stand and Carousel holds and stores pipets in an upright position to avoid contamination. For use with PICUS™ and eLINE™ mechanical and electronic pipets.
383.00 € - 882.00 €
Specifications
For Use With | Pipette Tips |
---|---|
Material | Polypropylene |
Type | Pipette Holder |
For Use With (Equipment) | Picus and eLINE Electronic Pipettes |
Includes | Universal AC adapter |
Includes: Universal AC-adaptor
Description
For storing and charging Picus and eLINE mechanical and electronic pipets when not in use. Pipets should be stored in an upright position in order to avoid contamination from desks and benches.Specifications
Pipette Tips | |
Pipette Holder | |
Universal AC adapter |
Polypropylene | |
Picus and eLINE Electronic Pipettes |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Sartorius Pipet Charging Stand and Carousel