Learn More
BRAND™ Pipet Tips, 2 to 200uL: Standard Retention
GREENER_CHOICE
Manufactured from oleamide- and diHEMDA-free high-purity polypropylene. Pipet Tips, 2 to 200μL: Standard Resolution are thin-walled for high translucency, easy ejection, and low waste. Graduations marked at 20 and 100μL for quick volume check.
Brand:  BRAND™ 732208
39.30 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15104917
  • 49.85 € / Pack of 480
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • High-purity polypropylene is oleamide- and diHEMDA-free
  • Nonsterile
  • All unopened packages are DNA-, RNase-, ATP- and endotoxin-free
  • Thin-walled for high translucency, easy ejection and low waste
  • Graduations at 20 and 100μL for quick volume check
  • Available either in bulk packages or in convenient, ecological rack-and-stack formats
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2 to 200 μL
Colorless
DNase, RNase, ATP, and Endotoxin Free
Universal
Yes
20, 100μL
Non-sterile
Autoclavable
TipBox
TipBox: 5 boxes, 96 each
Polypropylene