Learn More
Gilson™ PIPETMAN™ L Starter Kit
Increases productivity, security and comfort. PIPETMAN™ L starter kit contains three pipettes, conveniently packaged with PIPETMAN DIAMOND Tips and useful accessories.
Brand: Gilson™ F167350
982.30 EUR valid until 2025-02-28
Use promo code "24154" to get your promotional price.
This item is not returnable.
View return policy
Description
- Three PIPETMAN L Pipettes: P20L, P200L, and P1000L with Gilson certificates
- Two racks of certified-quality PIPETMAN DIAMOND Tips: D200 and D1000
- Three SINGLE™ pipette holders
- Three Plastic tip ejectors
- One Gilson Guide to Pipetting
- One Two-minute inspection poster
Specifications
Autoclavable | |
1 | |
Pipette Starter Kit |
Blue | |
Pipetman L | |
2 to 20 μL (P20L), 20 to 200 μL (P200L), 100 to 1000 μL (P1000L) |