missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ PIPETMAN™ Comfort Pipette Handles
GREENER_CHOICE

For use with PIPETMAN Classic, Neo, G and Neo Multichannel pipettes

70.05 € - 959.00 €

Specifications

Color Blue
Type Handle
For Use With (Equipment) PIPETMAN Classic, Neo, G and Neo Multichannel pipettes
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15394867
View Documents
Gilson™
F161486
3/Pk.
70.05 €
Pack of 3
In Stock
to see stock.
Add to basket
 
15777617
GREENER_CHOICE
View Documents
Gilson™
F161485
50/Pk.
959.00 €
Pack of 50
Estimated Shipment: 13-03-2025
to see stock.
Add to basket
 
Description

Description

  • Simple to use and requires no tools
  • Designed to help reduce pipetting fatigue
  • Grip to support daily pipetting activity
  • Aids in reducing the risk of Repetitive Strain Injuries
  • Autoclavable at 121°C (252°F), 1 atm for 20 min.
Specifications

Specifications

Blue
Handle
PIPETMAN Classic, Neo, G and Neo Multichannel pipettes
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.