missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ PIPETMAN™ G
GREENER_CHOICE

PIPETMAN™ G offers a low pipetting force and hours of comfort due to a redesigned piston assembly system.

Brand:  Gilson™ F144072

753.35 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.


 View more versions of this product

Product Code. 16353186

  • 793.00 € / Each
Estimated Shipment: 30-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • With a redesigned piston assembly system and high quality piston grease, PIPETMAN™ G offers hours of comfortable pipetting
  • PIPETMAN™ G relies on the legendary accuracy, precision, and reliability of PIPETMAN™ with improved pipetting comfort
  • Made of PVDF and stainless steel
  • Offers premium durability
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Declaration of Conformity
Metal Ejector
8
Manual Air Displacement Pipette
3 Years
12.87 in.
Yellow
Not Autoclavable
P8x200G
Pipetman G
20 to 200 μL
5.98 in.
1.54 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gilson™ PIPETMAN™ G > Volume: 20 to 200 μL; 8-Channel

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.