missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ PIPETMAX™

PIPETMAX™ is an automated pipetting solution for the efficient processing of high-throughput biological assays, all in a benchtop size that easily fits into any lab.

34025.00 € - 38695.00 €

Specifications

Product Type Automated Pipetter
Certifications/Compliance ISO 9001
Channels 8
Material Polypropylene, Glass
Volume (Metric) 1 to 1000 μL
View More Specs

Products 3
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
16636165
View Documents
Gilson™
32100001
Cover Cutouts
34025.00 €
Each
Estimated Shipment: 24-03-2025
to see stock.
Add to basket
 
16626165
View Documents
Gilson™
32100000
Standard Cover
37885.00 €
Each
Estimated Shipment: 24-03-2025
to see stock.
Add to basket
 
16646165
View Documents
Gilson™
32100002
External Safety Interlock
38695.00 €
Each
Estimated Shipment: 24-03-2025
to see stock.
Add to basket
 
Description

Description

PIPETMAX is built upon PIPETMAN™ technology, helping to reduce costly technical replicates and maximize reproducibility by eliminating inconsistencies due to common pipetting errors. The automated system frees up time devoted to tedious pipetting jobs that improve lab productivity and allows you get to results faster. Several pre-developed, validated protocols are available for download while custom protocols can be written to accommodate any workflow.

  • Hardware and software of the PIPETMAX are customizable to fit specific needs of the assay, allowing you to configure and customize runs to any reagent, kit, or protocol
  • 8-channel cassettes can be set to use 1 to 8 tips
  • With a volume range from 1 to 1000 μL, PIPETMAX is an ideal solution for qPCR, NGS reactions, and cell-based assays
  • Small footprint and easily fits on lab bench or under a fume hood
  • Easy to move to location most suited for the assays at hand
Specifications

Specifications

Automated Pipetter
8
1 to 1000 μL
96- and 384-well microplates, strip plates, and cell culture formats
TRILUTION™
50/60 Hz, 100/240 V AC, 24 V DC, 6.25 A, 150 W
6.25 A
ISO 9001
Polypropylene, Glass
Liquid Handling
Touchscreen tablet, or PC control via USB and TRILUTION™ micro software
Polypropylene
100/240 V
50/60 Hz
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.