Learn More
BRAND™ Pipette Jar
Aluminum pipette box with inner silicone coating for storage and sterilization
Brand:  BRAND™ 28930
70.33 EUR valid until 2024-12-31
Use promo code "21877" to get your promotional price.
 View more versions of this product
Product Code. 15673957
  • 85.85 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Store pipettes of different lengths
  • Sterilize pipettes of different lengths
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Aluminum
    Rectangular
    Silicone
    Sterile
    Aluminum
    Pipette Storage and Sterilization
    Pipette Box
    315 to 385 mm