missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terracycle™ Pipette Tip Boxes Zero Waste Box™
Recycle Pipette Tip Boxes using this Zero Waste Box system. Zero Waste Box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.
Brand: Terracycle™ CS180-L-IE
This item is not returnable.
View return policy
Description
Use this Zero Waste Box to recycle:
- Empty pipette tip boxes
Do not send in:
- Pipette tips, tubes and rigid lab plastics that are contaminated with materials considered hazardous or dangerous
- Autoclaved material
- Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) which presents a danger to the environment, or to people
- Batteries, pressurized canisters, broken glass and medical waste
Important:
- We do not collect any hazardous waste containing substances with toxic, reactive, corrosive, infectious, flammable, explosive properties or harmful to human health and the environment.
- Please make sure that you do not mix hazardous waste with non-hazardous waste.
Specifications
25 kg | |
Beige, Green, Red, White | |
Single-use | |
Large | |
Not Autoclavable | |
Recycling Box |
55 lb. | |
38 x 38 x 97 cm | |
Yes | |
Non-sterile | |
Cardboard, Plastic |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction