missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kartell™ Pipette Filler with Adapter
Kartell™ Pipette Filler with Adapter is compatable with bulb or cylindrical pipettes of various capacities and features no metal parts that could potentially corrode.
Brand: Kartell™ 0020000
Description
Kartell™ Pipette Filler with Adapter is compatable with bulb or cylindrical pipettes of various capacities and features no metal parts that could potentially corrode.
- Moulded red rubber bulb
- Included three glass ball valves which can be controlled by finger pressure
- There are no metal parts to corrode and the filler can be used with all liquids except those that attack rubber
- Can be used on bulb or cylindrical pipettes of different capacity
- Complete with adapter for pipettes with capacities more than 20mL
Specifications
Red | |
Ball | |
Adapter for pipettes with capacities greater than 20mL |
Rubber | |
20 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction