missing translation for 'onlineSavingsMsg'
Learn More

Caframo™ Pitched Blade Propellers, Blade Only

Wide selection of stainless steel pitched blade propellers for use with Caframo™ and Fisherbrand™ Overhead Stirrers and shafts

Brand:  Caframo™ A521

Product Code. 12997116

  • 58.55 € / Each
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Electro-polished stainless steel 316L
  • Chemically inert and corrosion resistant 316L stainless steel
  • For low shear, high top-to-bottom pumping capacity
  • Scale up advantage if propeller used in production

Order Info

Shafts sold separately

Specifications

Specifications

3.8 cm
Caframo™ Overhead Stirrer
8 mm Bore, Set Screw
Pitched Blade Propeller, Axial Flow
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.