missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Plain Interior Polypropylene Funnels
An excellent option with high temperature resistance for general laboratory use.
Brand: BRAND™ 147035
43.25 EUR valid until 2024-12-31
Use promo code "21745" to get your promotional price.
This item is not returnable.
View return policy
Description
- Available in a range of sizes
- Sold in various quantities
Specifications
150mm | |
Polypropylene | |
Autoclavable | |
12/Pk. |
17mm | |
95 mm (Stem) | |
95mm | |
Filling Funnel |