Learn More
Fisherbrand™ Plain PTFE Stir Bars
Provides stable stirring action. Fisherbrand™ Plain PTFE Stir Bars provide greater turbulence than cylindrical stir bars.
Brand:  Fisherbrand™ 001.260
 View more versions of this product
Product Code. 16215801
  • 35.65 € / Pack of 10
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Stable, general-purpose magnetic stir bar
  • Provides greater turbulence than cylindrical stir bars
  • Autoclavable
Specifications
Specifications
Autoclavable
White
Containers & Vessels / Stirrer Hot Plates
60 mm
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
PTFE encapsulated Alnico V magnet core
10 mm
2/Pk.