missing translation for 'onlineSavingsMsg'
Learn More

Plas Labs™ Flat-Bottom Rodent Restrainers

Apertures in the top and bottom provide easy access for injections or sampling

Specifications

Color Clear
For Use With (Application) Injections or Blood Sampling
Type Flat Bottom Rodent Restrainer
Material Noryl™ Plastic
Sterilization Method Chemically sterilized or washed at 180°F
View More Specs

Description

Description

  • Pocket and holes in the front make it easy to anesthetize or feed the animal
  • Clear material provides a visual check of rodent's condition
  • Adjustable tail stock can be inserted into any one of four positions

Specifications

Specifications

Clear
Flat Bottom Rodent Restrainer
Chemically sterilized or washed at 180°F
Injections or Blood Sampling
Noryl™ Plastic
Videos
SDS
Documents

Documents

Product Certifications