missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ PP/PE Plastic Barrel Funnels
All-purpose funnels with suspension eyelet for safe filling of containers.
Brand: Buerkle™ 9602-0250
This item is not returnable.
View return policy
Description
- Manufactured from either polypropylene or polyethylene
- Available in a range of diameters
Specifications
250 mm | |
Polypropylene | |
Suspension eyelet |
30 mm | |
For Foodstuffs | |
Barrel Funnel |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction