missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ PP/PE Plastic Barrel Funnels

All-purpose funnels with suspension eyelet for safe filling of containers.

Brand:  Buerkle™ 9602-0400

 View more versions of this product

Product Code. 11961491

  • 105.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Manufactured from either polypropylene or polyethylene
  • Available in a range of diameters
Specifications

Specifications

400 mm
Polyethylene
Suspension eyelet
42 mm
For Foodstuffs
Barrel Funnel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buerkle™ PP/PE Plastic Barrel Funnels > Top Diameter: 400mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.