missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Plastic Connector

Plastic Connectors 25 mm TC - Threaded

Brand:  Cytiva 18-1169-22

Product Code. 16467645

  • 170.00 € / Pack of 2
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Polypropylene
  • Connections: 25 mm TC i.d. 6 mm to 6 mm (1/4 in.) Threaded
TRUSTED_SUSTAINABILITY
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers