Learn More
Nickel Electro™ Plastic Flat Lid
Reduce the evaporation and noise in an Ultrasonic Bath. Nickel Electro™ Clifton™ Plastic Flat Lid only requires one piece construction.
Brand:  Nickel Electro™ LD-1
 View more versions of this product
Product Code. 13161323
  • 41.50 € / Each
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • One piece construction
  • Reduces evaporation
  • Reduces noise
  • For use with Ultrasonic Bath
Specifications
Specifications
Ultrasonic Bath SW1H
Plastic